Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002949.3 | NP_002940.2 | 2 | 358 | |
GenBank mRNA | AF105278.1 | - | 1 | 92 | |
BC002344.2 | - | 2 | 343 | 2961288 | |
BC007497.2 | - | 2 | 240 | 2959986 | |
DA648796.1 | - | 2 | 358 | ||
EF036488.1 | - | 2 | 228 | ||
U25041.1 | - | 2 | 244 | ||
X79865.1 | - | 2 | 363 |