Gene Transcripts

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 19: 40292743 - 40348527 on Build GRCh38
UniGene ID
Hs.631557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256440.1 NP_001243369.1 9 1172
NM_001256441.1 NP_001243370.1 9 1167
NR_046202.1 9 1220
XM_005258520.2 XP_005258577.1 9 1161
XM_017026291.1 XP_016881780.1 9 1172
XM_017026294.1 XP_016881783.1 6 890
XM_017026295.1 XP_016881784.1 10 1270
GenBank mRNA AK094207.1 - 9 1160
AL834131.1 - 9 1197
BC023623.2 - 9 1198 4662735
BC027935.1 - 9 1188 5202287
CR936760.1 - 9 1173

View Full Product Details