Gene Transcripts

Gene Symbol
WHRN
Entrez Gene ID
25861
Gene Name
whirlin
Gene Aliases
CIP98, DFNB31, PDZD7B, USH2D, WI
Chromosome Location
Chr. 9: 114402078 - 114505509 on Build GRCh38
UniGene ID
Hs.93836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083885.2 NP_001077354.2 5, 6 702
NM_001173425.1 NP_001166896.1 5, 6 1866
NM_015404.3 NP_056219.3 5, 6 1866
XM_011518484.1 XP_011516786.1 5, 6 1919
XM_011518485.1 XP_011516787.1 6, 7 1767
XM_011518486.1 XP_011516788.1 5, 6 1919
XM_011518487.1 XP_011516789.1 4, 5 1793
XM_011518488.1 XP_011516790.1 5, 6 1919
XM_011518489.2 XP_011516791.1 5, 6 1919
XM_011518491.2 XP_011516793.1 5, 6 1919
XM_011518492.1 XP_011516794.1 5, 6 1919
XM_011518494.2 XP_011516796.1 5, 6 1919
XM_011518495.1 XP_011516797.1 2, 3 171
GenBank mRNA AB040959.1 - 5, 6 1380
AK022854.1 - 1, 2 580
AL110228.1 - 5, 6 660
AL557526.3 - 1, 2 379
BC014524.1 - 2366
BC136416.1 - 5, 6 1615 9020404
BC142614.1 - 5, 6 1866 40147287
BC142684.1 - 5, 6 1841 40148465

View Full Product Details