Gene Transcripts

Gene Symbol
TEX19
Entrez Gene ID
400629
Gene Name
testis expressed 19
Gene Aliases
Chromosome Location
Chr. 17: 82359247 - 82363776 on Build GRCh38
UniGene ID
Hs.231897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207459.3 NP_997342.1 2 771
XM_017024638.1 XP_016880127.1 2 830
XM_017024639.1 XP_016880128.1 2 858
GenBank mRNA AK093086.1 - 2 774
BC016939.2 - 1 732 4419895
BC036191.1 - 2 771 5297531
BC050391.1 - 2 748 5745215
BC057820.1 - 2 772 4837469

View Full Product Details