Gene Transcripts

Gene Symbol
IRX1
Entrez Gene ID
79192
Gene Name
iroquois homeobox 1
Gene Aliases
IRX-5, IRXA1
Chromosome Location
Chr. 5: 3595855 - 3601403 on Build GRCh38
UniGene ID
Hs.424156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024337.3 NP_077313.3 4 1603
XM_017009845.1 XP_016865334.1 3 1729
GenBank mRNA AY335938.1 - 4 1606
BC029160.1 - 4 1305

View Full Product Details