Gene Transcripts

Gene Symbol
KRBA2
Entrez Gene ID
124751
Gene Name
KRAB-A domain containing 2
Gene Aliases
Chromosome Location
Chr. 17: 8368047 - 8376902 on Build GRCh38
UniGene ID
Hs.304604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304947.1 NP_001291876.1 2 1605
NM_213597.3 NP_998762.1 2 1025
XM_005256443.2 XP_005256500.1 5 2182
XM_011523643.1 XP_011521945.1 4 1998
XM_011523644.1 XP_011521946.1 3 1789
XM_017024163.1 XP_016879652.1 4 2271
XM_017024164.1 XP_016879653.1 2 1260
XM_017024165.1 XP_016879654.1 3 2085
GenBank mRNA AK131219.1 - 2 1026
BC033390.1 - 2 860

View Full Product Details