Gene Transcripts

Gene Symbol
PLA2G4F
Entrez Gene ID
255189
Gene Name
phospholipase A2 group IVF
Gene Aliases
PLA2G4FZ
Chromosome Location
Chr. 15: 42141134 - 42156641 on Build GRCh38
UniGene ID
Hs.231873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213600.3 NP_998765.3 3 293
NR_033151.1 2 220
GenBank mRNA AK122623.1 - 3 264
AK172836.1 - 3 293
BC035335.1 - 2 190
BC131531.1 - 3 270 40036909
BC142682.1 - 3 264 40148418
BC146648.1 - 3 264 40147152

View Full Product Details