Gene Transcripts

Gene Symbol
OR8D1
Entrez Gene ID
283159
Gene Name
olfactory receptor family 8 subfamily D member 1
Gene Aliases
JCG9, OR8D3, OST004, PDJ9J14
Chromosome Location
Chr. 11: 124309840 - 124310766 on Build GRCh38
UniGene ID
Hs.504211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002917.1 NP_001002917.1 1 916
GenBank mRNA AF238489.1 - 1 916
BC136950.1 - 1 987 9020942
BC136954.1 - 1 987 9020946

View Full Product Details