Gene Transcripts

Gene Symbol
OR51G2
Entrez Gene ID
81282
Gene Name
olfactory receptor family 51 subfamily G member 2
Gene Aliases
OR11-28
Chromosome Location
Chr. 11: 4914719 - 4915663 on Build GRCh38
UniGene ID
Hs.542302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005238.1 NP_001005238.1 1 486
GenBank mRNA BC136830.1 - 1 515
BC136831.1 - 1 515 9020822

View Full Product Details