Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081550.1 | NP_001075019.1 | 22 | 2425 | |
XM_005262450.3 | XP_005262507.1 | 22 | 2425 | ||
XM_017029662.1 | XP_016885151.1 | 23 | 2575 | ||
XM_017029663.1 | XP_016885152.1 | 23 | 2575 | ||
XM_017029664.1 | XP_016885153.1 | 23 | 2575 | ||
XM_017029665.1 | XP_016885154.1 | 23 | 2575 | ||
XM_017029666.1 | XP_016885155.1 | 23 | 2563 | ||
XM_017029667.1 | XP_016885156.1 | 23 | 2575 | ||
XM_017029668.1 | XP_016885157.1 | 23 | 2575 | ||
XM_017029669.1 | XP_016885158.1 | 23 | 2575 | ||
XM_017029670.1 | XP_016885159.1 | 23 | 2563 | ||
XM_017029671.1 | XP_016885160.1 | 23 | 2575 | ||
XM_017029672.1 | XP_016885161.1 | 23 | 2575 | ||
XM_017029673.1 | XP_016885162.1 | 22 | 2437 | ||
XM_017029674.1 | XP_016885163.1 | 22 | 2425 | ||
XM_017029675.1 | XP_016885164.1 | 23 | 2575 | ||
XM_017029676.1 | XP_016885165.1 | 23 | 2575 | ||
XM_017029677.1 | XP_016885166.1 | 22 | 2437 | ||
XM_017029678.1 | XP_016885167.1 | 22 | 2425 | ||
XM_017029679.1 | XP_016885168.1 | 22 | 2437 | ||
GenBank mRNA | AF441770.1 | - | 17 | 2048 | |
AK001758.1 | - | 20 | 2105 | ||
AK023659.1 | - | 7 | 719 | ||
AK296780.1 | - | 19 | 2197 | ||
BC063692.1 | - | 2 | 114 |