Gene Transcripts

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
Chromosome Location
Chr. 19: 13906143 - 13930880 on Build GRCh38
UniGene ID
Hs.269592
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017721.4 NP_060191.3 14 1843
XM_005259972.2 XP_005260029.1 14 1788
XM_005259973.2 XP_005260030.1 14 1791
XM_005259974.2 XP_005260031.1 14 1779
XM_005259975.2 XP_005260032.1 14 1776
GenBank mRNA AB097002.1 - 14 1785
AF536205.1 - 16 1896
AK000248.1 - 1602
AK023399.1 - 7 668
AK315068.1 - 14 1761
BC006556.2 - 9 947
BC064981.1 - 14 1747 6585236

View Full Product Details