Gene Transcripts
- Gene Symbol
- CEP112
- Entrez Gene ID
- 201134
- Gene Name
- centrosomal protein 112
- Gene Aliases
- CCDC46, MACOCO
- Chromosome Location
- Chr. 17: 65635540 - 66192131 on Build GRCh38
- UniGene ID
- Hs.408676
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037325.2 |
NP_001032402.1
|
6, 7 |
884 |
|
NM_001199165.2 |
NP_001186094.1
|
26, 27 |
3015 |
|
NM_001302891.1 |
NP_001289820.1
|
24, 25 |
2965 |
|
NR_126542.1 |
|
5, 6 |
714 |
|
XM_005257119.4 |
XP_005257176.1
|
26, 27 |
3109 |
|
XM_005257125.3 |
XP_005257182.1
|
18, 19 |
2099 |
|
XM_005257126.4 |
XP_005257183.1
|
15, 16 |
1764 |
|
XM_006721740.2 |
XP_006721803.1
|
26, 27 |
2981 |
|
XM_011524461.2 |
XP_011522763.1
|
26, 27 |
3122 |
|
XM_011524464.2 |
XP_011522766.1
|
24, 25 |
2898 |
|
XM_011524465.2 |
XP_011522767.1
|
23, 24 |
2438 |
|
XM_011524466.2 |
XP_011522768.1
|
16, 17 |
1917 |
|
XM_011524467.2 |
XP_011522769.1
|
15, 16 |
1759 |
|
XM_017024302.1 |
XP_016879791.1
|
24, 25 |
2852 |
|
GenBank mRNA |
AF458591.1 |
- |
16, 17 |
1922 |
|
AK098070.1 |
- |
16, 17 |
1834 |
|
AK299075.1 |
- |
26, 27 |
2966 |
|
BC029524.1 |
- |
6, 7 |
884 |
|
BC131805.1 |
- |
24, 25 |
2793 |
40119069 |
BG203878.1 |
- |
|
181 |
|
BG531260.1 |
- |
3, 4 |
202 |
|
BX097629.1 |
- |
6, 7 |
562 |
|
View Full Product Details