Gene Transcripts
- Gene Symbol
- TVP23B
- Entrez Gene ID
- 51030
- Gene Name
- trans-golgi network vesicle protein 23 homolog B (S. cerevisiae)
- Gene Aliases
- CGI-148, FAM18B, FAM18B1, NPD008, YDR084C
- Chromosome Location
- Chr. 17: 18780995 - 18806713 on Build GRCh38
- UniGene ID
- Hs.87295
- Species
- Homo sapiens
- Gene Symbol
- TVP23C
- Entrez Gene ID
- 201158
- Gene Name
- trans-golgi network vesicle protein 23 homolog C (S. cerevisiae)
- Gene Aliases
- FAM18B2
- Chromosome Location
- Chr. 17: 15502264 - 15563631 on Build GRCh38
- UniGene ID
- Hs.164595
- Species
- Homo sapiens
View Full Product Details