Gene Transcripts

Gene Symbol
KLHL30
Entrez Gene ID
377007
Gene Name
kelch like family member 30
Gene Aliases
Chromosome Location
Chr. 2: 238138722 - 238152906 on Build GRCh38
UniGene ID
Hs.591595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198582.3 NP_940984.3 8 3636
GenBank mRNA AK125364.1 - 7 3506

View Full Product Details