Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017369.2 | NP_001017369.1 | 5 | 786 | |
NM_006745.4 | NP_006736.1 | 6 | 1072 | ||
XM_005263176.2 | XP_005263233.1 | 6 | 1189 | ||
GenBank mRNA | AK295432.1 | - | 5 | 732 | |
BC010653.1 | - | 5 | 966 | 3866053 | |
BC107879.1 | - | 6 | 1034 | 6498540 | |
U60205.1 | - | 5 | 968 | ||
U93162.1 | - | 6 | 1032 |