Gene Transcripts

Gene Symbol
Tceanc
Entrez Gene ID
245695
Gene Name
transcription elongation factor A (SII) N-terminal and central domain containing
Gene Aliases
A630018P17Rik, Gm390
Chromosome Location
Chr. X: 166499812 - 166518598 on Build GRCm38
UniGene ID
Mm.336318
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007577.2 NP_001007578.2 5 1474
GenBank mRNA AK140929.1 - 5 1464
BC067008.1 - 5 1473

View Full Product Details