Gene Transcripts

Gene Symbol
Mpzl1
Entrez Gene ID
360871
Gene Name
myelin protein zero-like 1
Gene Aliases
Chromosome Location
Chr. 13: 83805244 - 83842066 on Build Rnor_6.0
UniGene ID
Rn.8087
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007728.1 NP_001007729.1 2 281
XM_006250180.2 XP_006250242.1 2 329
GenBank mRNA BC078916.1 - 2 281

View Full Product Details