Gene Transcripts

Gene Symbol
Btk
Entrez Gene ID
367901
Gene Name
Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
Chromosome Location
Chr. X: 105360922 - 105390580 on Build Rnor_6.0
UniGene ID
Rn.26996
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007798.1 NP_001007799.1 2 263
GenBank mRNA AY787790.1 - 2 263
FQ215667.1 - 406

View Full Product Details