Gene Transcripts

Gene Symbol
CST9
Entrez Gene ID
128822
Gene Name
cystatin 9
Gene Aliases
CLM, CTES7A
Chromosome Location
Chr. 20: 23602410 - 23605973 on Build GRCh38
UniGene ID
Hs.698020
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008693.2 NP_001008693.2 2 571
GenBank mRNA AF494536.1 - 2 474
BC137302.1 - 2 571 9021299
BC137303.1 - 2 571 9021300

View Full Product Details