Gene Transcripts

Gene Symbol
Whrn
Entrez Gene ID
73750
Gene Name
whirlin
Gene Aliases
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
Chromosome Location
Chr. 4: 63414909 - 63496078 on Build GRCm38
UniGene ID
Mm.300397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008791.2 NP_001008791.1 13 3344
NM_001008792.2 NP_001008792.1 12 3323
NM_001008793.2 NP_001008793.1 12 3308
NM_028640.2 NP_082916.1 12 3311
GenBank mRNA AK004110.1 - 2 41
AK122523.1 - 12 2439
AY739114.1 - 12 3271
AY739115.1 - 13 3304
AY739116.1 - 12 3283
AY739117.1 - 12 3268
AY739118.1 - 9 1920
AY739119.1 - 8 1887
AY739120.1 - 7 1674
AY739121.1 - 7 1792
AY739122.1 - 8 2124
BC150757.1 - 12 2668 9087668

View Full Product Details