Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008859.1 | NP_001008859.1 | 3 | 165 | |
XM_006244464.2 | XP_006244526.1 | 4 | 314 | ||
XM_006244465.2 | XP_006244527.1 | 4 | 311 | ||
GenBank mRNA | AY310148.1 | - | 3 | 165 |