Gene Transcripts

Gene Symbol
Vom1r2
Entrez Gene ID
494253
Gene Name
vomeronasal 1 receptor 2
Gene Aliases
V1re26
Chromosome Location
Chr. 1: 86362432 - 86363340 on Build Rnor_6.0
UniGene ID
Rn.129463
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008903.1 NP_001008903.1 1 192
GenBank mRNA AY510349.1 - 1 192

View Full Product Details