Gene Transcripts

Gene Symbol
Vom1r36
Entrez Gene ID
494264
Gene Name
vomeronasal 1 receptor 36
Gene Aliases
V1rd26
Chromosome Location
Chr. 1: 72183354 - 72184265 on Build Rnor_6.0
UniGene ID
Rn.129429
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008913.1 NP_001008913.1 1 376
GenBank mRNA AY510335.1 - 1 376

View Full Product Details