Gene Transcripts

Gene Symbol
Vom1r56
Entrez Gene ID
494277
Gene Name
vomeronasal 1 receptor 56
Gene Aliases
V1rj2, V1rj5
Chromosome Location
Chr. 1: 66663965 - 66664921 on Build Rnor_6.0
UniGene ID
Rn.129412
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008929.1 NP_001008929.1 1 533
GenBank mRNA AY510280.1 - 1 533

Gene Symbol
LOC100909792
Entrez Gene ID
100909792
Gene Name
vomeronasal type-1 receptor 2-like
Gene Aliases
Chromosome Location
Chr. 2: 168228046 - 168229055 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749304.2 XP_003749352.1 1 586

View Full Product Details