Gene Transcripts

Gene Symbol
Vom1r52
Entrez Gene ID
494278
Gene Name
vomeronasal 1 receptor 52
Gene Aliases
V1rm6
Chromosome Location
Chr. 1: 66774431 - 66775384 on Build Rnor_6.0
UniGene ID
Rn.129415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008930.1 NP_001008930.1 1 930
GenBank mRNA AY510373.1 - 1 930

View Full Product Details