Gene Transcripts

Gene Symbol
Oas3
Entrez Gene ID
494202
Gene Name
2'-5'-oligoadenylate synthetase 3
Gene Aliases
Chromosome Location
Chr. 12: 41313081 - 41368217 on Build Rnor_6.0
UniGene ID
Rn.162137
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009493.1 NP_001009493.1 1168
GenBank mRNA AY250706.1 - 1168

View Full Product Details