Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 13 | 1252 | |
NM_012233.2 | NP_036365.1 | 13 | 1252 | ||
XM_011510823.2 | XP_011509125.1 | 13 | 1252 | ||
XM_011510825.2 | XP_011509127.1 | 13 | 1252 | ||
GenBank mRNA | AB449877.1 | - | 13 | 1209 | |
AK022494.1 | - | 13 | 1224 | ||
BC146809.1 | - | 15 | 1489 | 8860368 | |
D31886.1 | - | 13 | 1211 |