Gene Transcripts

Gene Symbol
Eln
Entrez Gene ID
25043
Gene Name
elastin
Gene Aliases
RATTREL11, TREL11, Trela, Trela26
Chromosome Location
Chr. 12: 24978478 - 25021864 on Build Rnor_6.0
UniGene ID
Rn.54384
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012722.1 NP_036854.1 37 3220
GenBank mRNA BC085910.1 - 37 3220
J04035.1 - 6 842

View Full Product Details