Gene Transcripts

Gene Symbol
PLEKHG4
Entrez Gene ID
25894
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Gene Aliases
ARHGEF44, PRTPHN1, SCA4
Chromosome Location
Chr. 16: 67277820 - 67289500 on Build GRCh38
UniGene ID
Hs.188781
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129727.2 NP_001123199.1 11 1616
NM_001129728.1 NP_001123200.1 10 1576
NM_001129729.2 NP_001123201.1 10 1549
NM_001129731.2 NP_001123203.1 8 1555
XM_011522985.2 XP_011521287.1 10 1546
XM_011522986.2 XP_011521288.1 10 1678
XM_011522987.2 XP_011521289.1 11 1622
XM_011522988.2 XP_011521290.1 10 1495
GenBank mRNA AB197663.1 - 9 3812
AB197664.1 - 8 3979
AK024475.1 - 10 1511
AK304723.1 - 8 967
AK308298.1 - 10 1485
AL117435.1 - 5 448
BC054486.1 - 8 1490 4820741
BC063501.1 - 10 1487 5527705
BC082974.1 - 11 1584 6291175
BC115390.1 - 7 1514

View Full Product Details