Gene Transcripts
- Gene Symbol
- L3MBTL3
- Entrez Gene ID
- 84456
- Gene Name
- l(3)mbt-like 3 (Drosophila)
- Gene Aliases
- MBT-1, MBT1
- Chromosome Location
- Chr. 6: 130018583 - 130141449 on Build GRCh38
- UniGene ID
- Hs.658051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007102.2 |
NP_001007103.1
|
3, 4 |
272 |
|
NM_032438.2 |
NP_115814.1
|
3, 4 |
272 |
|
XM_005267161.4 |
XP_005267218.1
|
3, 4 |
448 |
|
XM_006715576.3 |
XP_006715639.1
|
3, 4 |
272 |
|
XM_006715578.3 |
XP_006715641.1
|
3, 4 |
256 |
|
XM_011536179.2 |
XP_011534481.1
|
3, 4 |
255 |
|
XM_011536180.2 |
XP_011534482.1
|
3, 4 |
274 |
|
XM_011536181.2 |
XP_011534483.1
|
3, 4 |
209 |
|
XM_011536183.2 |
XP_011534485.1
|
3, 4 |
805 |
|
XM_011536184.2 |
XP_011534486.1
|
3, 4 |
209 |
|
XM_017011355.1 |
XP_016866844.1
|
3, 4 |
363 |
|
XM_017011356.1 |
XP_016866845.1
|
3, 4 |
255 |
|
GenBank mRNA |
AB058701.1 |
- |
2, 3 |
170 |
|
AK094137.1 |
- |
3, 4 |
278 |
|
AK290219.1 |
- |
3, 4 |
264 |
|
BC060845.1 |
- |
3, 4 |
266 |
30342272 |
DA927626.1 |
- |
3, 4 |
272 |
|
View Full Product Details