Gene Transcripts

Gene Symbol
Rasl11a
Entrez Gene ID
304268
Gene Name
RAS-like family 11 member A
Gene Aliases
Chromosome Location
Chr. 12: 9986043 - 9990284 on Build Rnor_6.0
UniGene ID
Rn.105901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002829.1 NP_001002829.1 6 593
XM_006248834.2 XP_006248896.1 4 1056

View Full Product Details