Gene Transcripts

Gene Symbol
Ano7
Entrez Gene ID
367318
Gene Name
anoctamin 7
Gene Aliases
Ngep, Tmem16g
Chromosome Location
Chr. 9: 100509660 - 100553261 on Build Rnor_6.0
UniGene ID
Rn.163084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004071.1 NP_001004071.1 8 811
XM_008767382.1 XP_008765604.1 9 2465

View Full Product Details