Gene Transcripts

Gene Symbol
Klk4
Entrez Gene ID
408210
Gene Name
kallikrein-related peptidase 4
Gene Aliases
Emsp1
Chromosome Location
Chr. 1: 99828944 - 99834642 on Build Rnor_6.0
UniGene ID
Rn.142022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004101.1 NP_001004101.1 3 377
XM_008759403.1 XP_008757625.1 4 419

View Full Product Details