Gene Transcripts

Gene Symbol
Rragb
Entrez Gene ID
245670
Gene Name
Ras-related GTP binding B
Gene Aliases
Chromosome Location
Chr. X: 153139796 - 153190192 on Build GRCm38
UniGene ID
Mm.190922
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004154.2 NP_001004154.1 4 806
GenBank mRNA BC069180.1 - 4 764
BC070407.1 - 4 783 6808979

View Full Product Details