Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014784.3 | NP_055599.1 | 7 | 1612 | |
NM_198236.2 | NP_937879.1 | 7 | 1612 | ||
XM_005245629.4 | XP_005245686.1 | 7 | 2013 | ||
XM_006711659.2 | XP_006711722.1 | 9 | 2061 | ||
XM_006711660.3 | XP_006711723.1 | 9 | 2061 | ||
XM_006711661.2 | XP_006711724.1 | 9 | 2061 | ||
XM_006711662.2 | XP_006711725.1 | 9 | 2061 | ||
XM_006711663.3 | XP_006711726.1 | 7 | 1048 | ||
XM_006711665.3 | XP_006711728.1 | 9 | 2061 | ||
XM_011510185.2 | XP_011508487.1 | 8 | 2046 | ||
XM_011510186.2 | XP_011508488.1 | 9 | 2061 | ||
XM_011510187.2 | XP_011508489.1 | 8 | 772 | ||
XM_017002920.1 | XP_016858409.1 | 8 | 2028 | ||
XM_017002921.1 | XP_016858410.1 | 7 | 2013 | ||
XM_017002922.1 | XP_016858411.1 | 9 | 2061 | ||
XM_017002923.1 | XP_016858412.1 | 7 | 2013 | ||
XM_017002924.1 | XP_016858413.1 | 8 | 2046 | ||
GenBank mRNA | AB002378.1 | - | 7 | 1317 | |
BC057394.1 | - | 7 | 1636 | 6148684 |