Gene Transcripts
- Gene Symbol
- TBC1D4
- Entrez Gene ID
- 9882
- Gene Name
- TBC1 domain family member 4
- Gene Aliases
- AS160, NIDDM5
- Chromosome Location
- Chr. 13: 75284664 - 75482646 on Build GRCh38
- UniGene ID
- Hs.210891
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286658.2 |
NP_001273587.1
|
1, 2 |
907 |
|
NM_001286659.2 |
NP_001273588.1
|
1, 2 |
907 |
|
NM_014832.4 |
NP_055647.2
|
1, 2 |
907 |
|
XM_005266603.1 |
XP_005266660.1
|
1, 2 |
1385 |
|
XM_011535331.1 |
XP_011533633.1
|
1, 2 |
1385 |
|
GenBank mRNA |
AB011175.1 |
- |
1, 2 |
853 |
|
AB449885.1 |
- |
1, 2 |
506 |
|
AK307493.1 |
- |
1, 2 |
907 |
|
BC151239.1 |
- |
1, 2 |
845 |
8860512 |
FM207106.1 |
- |
1, 2 |
506 |
|
FM207107.1 |
- |
1, 2 |
506 |
|
View Full Product Details