Gene Transcripts

Gene Symbol
OR10K1
Entrez Gene ID
391109
Gene Name
olfactory receptor family 10 subfamily K member 1
Gene Aliases
OR1-6
Chromosome Location
Chr. 1: 158465562 - 158466503 on Build GRCh38
UniGene ID
Hs.690410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004473.1 NP_001004473.1 1 893
GenBank mRNA BC137323.1 - 1 973
BC137324.1 - 1 973 9021321

View Full Product Details