Gene Transcripts

Gene Symbol
OR13F1
Entrez Gene ID
138805
Gene Name
olfactory receptor family 13 subfamily F member 1
Gene Aliases
OR9-6
Chromosome Location
Chr. 9: 104504263 - 104505222 on Build GRCh38
UniGene ID
Hs.553602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004485.1 NP_001004485.1 1 791
GenBank mRNA BC140739.1 - 1 880

View Full Product Details