Gene Transcripts

Gene Symbol
OR52L1
Entrez Gene ID
338751
Gene Name
olfactory receptor family 52 subfamily L member 1
Gene Aliases
OR11-50
Chromosome Location
Chr. 11: 5985892 - 5986985 on Build GRCh38
UniGene ID
Hs.553703
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005173.3 NP_001005173.3 1 387
GenBank mRNA BC137343.1 - 1 387

View Full Product Details