Gene Transcripts
- Gene Symbol
- FRG2
- Entrez Gene ID
- 448831
- Gene Name
- FSHD region gene 2
- Gene Aliases
- FRG2A
- Chromosome Location
- Chr. 4: 190024367 - 190027257 on Build GRCh38
- UniGene ID
- Hs.626907
- Species
- Homo sapiens
- Gene Symbol
- FRG2C
- Entrez Gene ID
- 100288801
- Gene Name
- FSHD region gene 2 family member C
- Gene Aliases
- Chromosome Location
- Chr. 3: 75646761 - 75667217 on Build GRCh38
- UniGene ID
- Hs.274541
- Species
- Homo sapiens
- Gene Symbol
- FRG2EP
- Entrez Gene ID
- 647476
- Gene Name
- FSHD region gene 2 family member E, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 20: 29414075 - 29416961 on Build GRCh38
- UniGene ID
- Hs.274541
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037925.1 |
|
4 |
1618 |
|
GenBank mRNA |
AK000525.1 |
- |
4 |
1618 |
|
View Full Product Details