Gene Transcripts
- Gene Symbol
- EBF3
- Entrez Gene ID
- 253738
- Gene Name
- early B-cell factor 3
- Gene Aliases
- COE3, EBF-3, O/E-2, OE-2
- Chromosome Location
- Chr. 10: 129835232 - 129964274 on Build GRCh38
- UniGene ID
- Hs.591374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005463.2 |
NP_001005463.1
|
2, 3 |
361 |
|
XM_005252667.3 |
XP_005252724.1
|
2, 3 |
808 |
|
XM_005252668.3 |
XP_005252725.1
|
2, 3 |
808 |
|
XM_005252669.3 |
XP_005252726.1
|
2, 3 |
808 |
|
XM_006717739.3 |
XP_006717802.1
|
2, 3 |
808 |
|
XM_006717740.3 |
XP_006717803.1
|
2, 3 |
808 |
|
XM_006717741.3 |
XP_006717804.1
|
2, 3 |
808 |
|
XM_006717742.3 |
XP_006717805.1
|
2, 3 |
808 |
|
XM_006717743.3 |
XP_006717806.1
|
2, 3 |
808 |
|
XM_006717744.3 |
XP_006717807.1
|
2, 3 |
808 |
|
XM_011539574.2 |
XP_011537876.1
|
2, 3 |
808 |
|
XM_017016027.1 |
XP_016871516.1
|
2, 3 |
808 |
|
GenBank mRNA |
AK097335.1 |
- |
3 |
771 |
|
AL832828.1 |
- |
2, 3 |
288 |
|
BC126130.1 |
- |
2, 3 |
352 |
8991846 |
BC130479.1 |
- |
2, 3 |
352 |
40146510 |
View Full Product Details