Gene Transcripts

Gene Symbol
LPGAT1
Entrez Gene ID
9926
Gene Name
lysophosphatidylglycerol acyltransferase 1
Gene Aliases
FAM34A, FAM34A1, NET8
Chromosome Location
Chr. 1: 211743457 - 211830772 on Build GRCh38
UniGene ID
Hs.744851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320808.1 NP_001307737.1 2 999
NM_014873.2 NP_055688.1 2 410
XM_005273362.3 XP_005273419.1 3 558
XM_011510228.2 XP_011508530.1 3 447
XM_011510229.2 XP_011508531.1 2 330
GenBank mRNA AY561706.1 - 1 183
BC034621.1 - 2 1000 4839400
BP369253.1 - 2 294
D86960.1 - 2 410

View Full Product Details