Gene Transcripts
- Gene Symbol
- JOSD1
- Entrez Gene ID
- 9929
- Gene Name
- Josephin domain containing 1
- Gene Aliases
- dJ508I15.2
- Chromosome Location
- Chr. 22: 38685543 - 38701418 on Build GRCh38
- UniGene ID
- Hs.3094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014876.5 |
NP_055691.1
|
1, 2 |
660 |
|
XM_005261876.2 |
XP_005261933.1
|
2, 3 |
967 |
|
XM_005261877.2 |
XP_005261934.1
|
2, 3 |
863 |
|
XM_005261878.2 |
XP_005261935.1
|
2, 3 |
891 |
|
XM_005261879.2 |
XP_005261936.1
|
2, 3 |
986 |
|
GenBank mRNA |
AK291827.1 |
- |
1, 2 |
453 |
|
BC015026.2 |
- |
1, 2 |
494 |
3921785 |
CR456473.1 |
- |
1, 2 |
339 |
|
CR456923.1 |
- |
1, 2 |
193 |
|
D31884.1 |
- |
1, 2 |
472 |
|
View Full Product Details