Gene Transcripts

Gene Symbol
Srfbp1
Entrez Gene ID
291469
Gene Name
serum response factor binding protein 1
Gene Aliases
p49/STRAP
Chromosome Location
Chr. 18: 47456406 - 47478532 on Build Rnor_6.0
UniGene ID
Rn.101802
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005536.1 NP_001005536.1 6 536
XM_008772109.1 XP_008770331.1 4 417
GenBank mRNA BC082077.1 - 6 536

View Full Product Details