Gene Transcripts

Gene Symbol
NSA2
Entrez Gene ID
10412
Gene Name
NSA2, ribosome biogenesis homolog
Gene Aliases
CDK105, HCL-G1, HCLG1, HUSSY-29, HUSSY29, TINP1
Chromosome Location
Chr. 5: 74765429 - 74776912 on Build GRCh38
UniGene ID
Hs.482526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271665.1 NP_001258594.1 2 459
NM_014886.4 NP_055701.1 2 459
NR_073403.1 2 459
XM_011543098.1 XP_011541400.1 2 137
XM_017008952.1 XP_016864441.1 2 137
XM_017008953.1 XP_016864442.1 2 209
GenBank mRNA AF077615.1 - 2 173
AF372458.1 - 2 184
AJ012409.1 - 2 171
AK313784.1 - 2 202
AY443419.1 - 1 89
BC005288.1 - 2 202 3683720
HY021659.1 - 2 460

View Full Product Details