Gene Transcripts

Gene Symbol
Pi4k2b
Entrez Gene ID
305419
Gene Name
phosphatidylinositol 4-kinase type 2 beta
Gene Aliases
Chromosome Location
Chr. 14: 60567968 - 60594821 on Build Rnor_6.0
UniGene ID
Rn.21189
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005883.1 NP_001005883.1 2 392
GenBank mRNA BC083668.1 - 2 392

View Full Product Details