Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164232.1 | NP_001157704.1 | 18 | 3170 | |
NM_015214.2 | NP_056029.2 | 18 | 3347 | ||
XM_005273455.3 | XP_005273512.1 | 18 | 3531 | ||
XM_005273456.3 | XP_005273513.1 | 17 | 3405 | ||
XM_017013252.1 | XP_016868741.1 | 18 | 3304 | ||
XM_017013254.1 | XP_016868743.1 | 17 | 3084 | ||
XM_017013255.1 | XP_016868744.1 | 16 | 3093 | ||
GenBank mRNA | AB018268.1 | - | 15 | 2557 | |
AK023218.1 | - | 14 | 2376 | ||
AK026961.1 | - | 3 | 968 | ||
AK056525.1 | - | 18 | 3249 | ||
AK125904.1 | - | 18 | 3170 | ||
AK127040.1 | - | 9 | 2698 |