Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242307.1 | NP_001229236.1 | 14 | 1678 | |
NM_001242309.1 | NP_001229238.1 | 13 | 1575 | ||
NM_014889.3 | NP_055704.2 | 14 | 1678 | ||
XM_005252345.3 | XP_005252402.1 | 14 | 1678 | ||
XM_017015470.1 | XP_016870959.1 | 14 | 1746 | ||
XM_017015471.1 | XP_016870960.1 | 14 | 1746 | ||
XM_017015472.1 | XP_016870961.1 | 14 | 1575 | ||
XM_017015473.1 | XP_016870962.1 | 14 | 1575 | ||
GenBank mRNA | AB029027.2 | - | 4 | 407 | |
AF061243.1 | - | 14 | 1622 | ||
AK002061.1 | - | 14 | 1646 | ||
AK023476.1 | - | 7 | 793 | ||
AK225084.1 | - | 14 | 1616 | ||
AK294865.1 | - | 13 | 1591 | ||
AK299464.1 | - | 13 | 1424 | ||
AK303406.1 | - | 13 | 1516 | ||
AK315117.1 | - | 14 | 1616 | ||
BC001150.1 | - | 8 | 978 | 3503609 | |
BC005025.1 | - | 14 | 1613 | 4040225 | |
BC045577.1 | - | 14 | 2088 | ||
BC095422.1 | - | 14 | 1619 | 4822631 | |
BC111987.1 | - | 14 | 1634 | 8327455 | |
BC113369.1 | - | 14 | 1634 | 8322421 | |
BX649017.1 | - | 14 | 1620 | ||
CR749279.1 | - | 12 | 1354 |