Gene Transcripts

Gene Symbol
Ier3
Entrez Gene ID
294235
Gene Name
immediate early response 3
Gene Aliases
Dif2, Iex1, Ler3, Prg1
Chromosome Location
Chr. 20: 3438798 - 3440002 on Build Rnor_6.0
UniGene ID
Rn.23638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212505.2 NP_997670.1 2 710
GenBank mRNA BC099831.1 - 2 686

View Full Product Details