Gene Transcripts

Gene Symbol
Olr16
Entrez Gene ID
404899
Gene Name
olfactory receptor 16
Gene Aliases
Chromosome Location
Chr. 1: 149674493 - 149675443 on Build Rnor_6.0
UniGene ID
Rn.141951
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000694.1 NP_001000694.1 890

Gene Symbol
LOC100910009
Entrez Gene ID
100910009
Gene Name
olfactory receptor 14A2-like
Gene Aliases
Chromosome Location
Chr. 1: 149858460 - 149859555 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748917.2 XP_003748965.1 1 966

Gene Symbol
Olr294
Entrez Gene ID
100909894
Gene Name
olfactory receptor 294
Gene Aliases
Chromosome Location
Chr. 1: 149769762 - 149770857 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748914.2 XP_003748962.1 966

View Full Product Details